Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRMS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192544.1 | NP_001179473.1 | 6 - 7 | 1132 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Bos taurus Probe spans exons |