Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCF20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192574.2 | NP_001179503.2 | 3 - 4 | 5823 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |