Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFNB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206283.2 | NP_001193212.2 | 4 - 5 | 986 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Bos taurus Probe spans exons |