Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEX3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206290.2 | NP_001193219.2 | 1 - 2 | 457 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Bos taurus Probe spans exons |