Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MZT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206331.1 | NP_001193260.1 | 1 - 2 | 111 | 106 | |
GenBank mRNA | DN549513.1 | - | 1 - 2 | 81 | 106 | |
DT834560.1 | - | 1 - 2 | 112 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Bos taurus Probe spans exons |