Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206415.1 | NP_001193344.1 | 1 - 2 | 200 | 69 | |
GenBank mRNA | DV828559.1 | - | 1 - 2 | 200 | 69 | |
DV886299.1 | - | 1 - 2 | 136 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Bos taurus Probe spans exons |