Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206428.1 | NP_001193357.1 | 4 - 5 | 1149 | 87 | |
GenBank mRNA | BC149548.1 | - | 4 - 5 | 1150 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |