Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BIVM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206453.2 | NP_001193382.2 | 4 - 5 | 1208 | 104 | |
GenBank mRNA | DV823208.1 | - | 3 - 4 | 280 | 104 | |
EE907980.1 | - | 2 - 3 | 211 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Bos taurus Probe spans exons |