Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPPA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206481.2 | NP_001193410.1 | 5 - 6 | 721 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |