Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMGCS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206578.1 | NP_001193507.1 | 3 - 4 | 666 | 83 | |
GenBank mRNA | BC102850.1 | - | 3 - 4 | 576 | 83 | |
DT814755.1 | - | 3 - 4 | 607 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |