Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HCRTR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192677.1 | NP_001179606.1 | 6 - 7 | 1104 | 80 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Bos taurus Probe spans exons |