Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QRFPR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192681.1 | NP_001179610.1 | 2 - 3 | 857 | 87 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Bos taurus Probe spans exons |