Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNA15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192714.2 | NP_001179643.1 | 3 - 4 | 850 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Bos taurus Probe spans exons |