Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192749.1 | NP_001179678.1 | 1 - 2 | 153 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |