Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABRB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192760.2 | NP_001179689.2 | 10 - 11 | 1207 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Bos taurus Probe spans exons |