Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CGRRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192807.1 | NP_001179736.1 | 5 - 6 | 761 | 122 | |
GenBank mRNA | BC122778.1 | - | 5 - 6 | 755 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |