Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163943.1 | NP_001157415.1 | 3 - 4 | 303 | 126 | |
GenBank mRNA | BC103345.1 | - | 3 - 4 | 303 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |