Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163946.2 | NP_001157418.1 | 5 - 6 | 554 | 81 | |
GenBank mRNA | BC149694.1 | - | 5 - 6 | 555 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |