Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OVOS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164024.1 | NP_001157496.1 | 19 - 20 | 2700 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |