Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIPM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206597.1 | NP_001193526.1 | 1 - 2 | 146 | 79 | |
GenBank mRNA | DV917166.1 | - | 2 - 3 | 128 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |