Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIPM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206597.1 | NP_001193526.1 | 7 - 8 | 933 | 70 | |
GenBank mRNA | DV916511.1 | - | 5 - 6 | 623 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |