Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206633.1 | NP_001193562.1 | 2 - 3 | 219 | 58 | |
GenBank mRNA | AY397646.1 | - | 1 - 2 | 117 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Bos taurus Inventoried Probe spans exons |