Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206687.2 | NP_001193616.1 | 2 - 3 | 154 | 60 | |
GenBank mRNA | DT860082.1 | - | 2 - 3 | 154 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |