Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206699.2 | NP_001193628.2 | 6 - 7 | 1603 | 87 | |
GenBank mRNA | EH376280.1 | - | 6 - 7 | 613 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |