Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206727.1 | NP_001193656.1 | 4 - 5 | 501 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |