Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR4D9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206742.1 | NP_001193671.1 | 1 | 617 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Bos taurus Both primers and probe map within a single exon |