Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBXN7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206764.2 | NP_001193693.1 | 5 - 6 | 495 | 65 | |
GenBank mRNA | EE353771.1 | - | 4 - 5 | 420 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |