Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTFMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166523.3 | NP_001159995.1 | 2 - 3 | 445 | 70 | |
GenBank mRNA | AB004316.1 | - | 2 - 3 | 380 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Bos taurus Probe spans exons |