Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166605.1 | NP_001160077.1 | 2 - 3 | 137 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |