Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC8B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177596.1 | NP_001171067.1 | 16 - 17 | 2143 | 76 | |
GenBank mRNA | BT026307.1 | - | 10 - 11 | 1137 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Bos taurus Probe spans exons |