Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSMB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190300.2 | NP_001177229.1 | 3 - 4 | 338 | 87 | |
GenBank mRNA | BC108152.1 | - | 3 - 4 | 339 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |