Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BEND5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190953.1 | NP_001177882.1 | 3 - 4 | 768 | 105 | |
GenBank mRNA | BC116004.1 | - | 3 - 4 | 697 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |