Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNNM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191172.1 | NP_001178101.1 | 3 - 4 | 1898 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |