Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191187.3 | NP_001178116.3 | 22 - 23 | 3026 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |