Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NME5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191216.2 | NP_001178145.2 | 1 - 2 | 130 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Bos taurus Probe spans exons |