Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NME5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191216.2 | NP_001178145.2 | 3 - 4 | 435 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |