Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191237.2 | NP_001178166.2 | 2 - 3 | 245 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Bos taurus Probe spans exons |