Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLGN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191242.1 | NP_001178171.1 | 4 - 5 | 850 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Bos taurus Probe spans exons |