Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRDM14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191264.1 | NP_001178193.1 | 6 - 7 | 1492 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Bos taurus Probe spans exons |