Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AJAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206815.1 | NP_001193744.1 | 2 - 3 | 952 | 101 | |
GenBank mRNA | EV704037.1 | - | 4 - 5 | 505 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |