Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206831.1 | NP_001193760.1 | 2 - 3 | 794 | 65 | |
GenBank mRNA | DV851442.1 | - | 2 - 3 | 733 | 65 | |
EH177301.1 | - | 2 - 3 | 277 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |