Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206853.1 | NP_001193782.1 | 7 - 8 | 1590 | 118 | |
GenBank mRNA | BC149309.1 | - | 5 - 6 | 856 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |