Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206939.1 | NP_001193868.1 | 2 - 3 | 771 | 67 | |
GenBank mRNA | DV924764.1 | - | 2 - 3 | 653 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Bos taurus Probe spans exons |