Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AOPEP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206980.2 | NP_001193909.2 | 1 - 2 | 39 | 96 | |
GenBank mRNA | DY049760.1 | - | 1 - 2 | 40 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |