Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC20B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206982.1 | NP_001193911.1 | 8 - 9 | 1138 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |