Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPACA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001207045.1 | NP_001193974.1 | 6 - 7 | 579 | 75 | |
GenBank mRNA | BC108171.1 | - | 6 - 7 | 580 | 75 | |
DT848094.1 | - | 6 - 7 | 549 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |