Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C18H19orf33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001207046.1 | NP_001193975.1 | 3 - 4 | 307 | 60 | |
GenBank mRNA | BC102086.1 | - | 3 - 4 | 308 | 60 | |
EV616576.1 | - | 3 - 4 | 258 | 60 | ||
EV688939.1 | - | 3 - 4 | 212 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |