Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF19A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191331.1 | NP_001178260.1 | 6 - 7 | 1647 | 102 | |
GenBank mRNA | BC151661.1 | - | 6 - 7 | 1626 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |