Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191339.2 | NP_001178268.2 | 3 - 4 | 539 | 87 | |
GenBank mRNA | BC149915.1 | - | 1 - 2 | 52 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |