Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCL6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191366.2 | NP_001178295.2 | 6 - 7 | 1096 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Bos taurus Probe spans exons |